Search
Search

MTRR| Gene Symbol: MTRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556580.2 | XP_005556637.1 | 9 - 10 | 1409 | 76 | |
| XM_005556581.1 | XP_005556638.1 | 10 - 11 | 1582 | 76 | ||
| XM_005556582.2 | XP_005556639.1 | 10 - 11 | 1557 | 76 | ||
| XM_005556583.1 | XP_005556640.1 | 9 - 10 | 1459 | 76 | ||
| XM_015451206.1 | XP_015306692.1 | 9 - 10 | 1792 | 76 | ||
| XM_015451207.1 | XP_015306693.1 | 9 - 10 | 1792 | 76 | ||
Set Membership: |
|