Search
Search

TRIM14| Gene Symbol: TRIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581219.2 | XP_005581276.1 | 5 - 6 | 1317 | 92 | |
| XM_015436952.1 | XP_015292438.1 | 5 - 6 | 991 | 92 | ||
Set Membership: |
|