Search
Search

PKD1L3| Gene Symbol: PKD1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442894.1 | XP_015298380.1 | 17 - 18 | 3050 | 54 | |
| XM_015442895.1 | XP_015298381.1 | 17 - 18 | 3050 | 54 | ||
| XM_015442896.1 | XP_015298382.1 | 17 - 18 | 3050 | 54 | ||
Set Membership: |
|