Search
Search

SERPINF2| Gene Symbol: SERPINF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582451.2 | XP_005582508.2 | 6 - 7 | 464 | 112 | |
| XM_005582452.2 | XP_005582509.2 | 5 - 6 | 475 | 112 | ||
| XM_015437436.1 | XP_015292922.1 | 5 - 6 | 396 | 112 | ||
Set Membership: |
|