Search
Search

LEMD1| Gene Symbol: LEMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540581.2 | XP_005540638.1 | 2 - 3 | 219 | 81 | |
| XM_005540582.2 | XP_005540639.1 | 2 - 3 | 236 | 81 | ||
| XM_005540583.2 | XP_005540640.1 | 2 - 3 | 219 | 81 | ||
| XM_005540584.2 | XP_005540641.1 | 2 - 3 | 219 | 81 | ||
| XM_005540588.2 | XP_005540645.1 | 2 - 3 | 219 | 81 | ||
| XM_015448928.1 | XP_015304414.1 | 2 - 3 | 219 | 81 | ||
Set Membership: |
|