Search
Search

LOC102122363| Gene Symbol: LOC102122363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562558.2 | XP_005562615.1 | 4 - 5 | 733 | 97 | |
| XM_005562559.2 | XP_005562616.1 | 4 - 5 | 481 | 97 | ||
| XM_005562560.2 | XP_005562617.1 | 4 - 5 | 559 | 97 | ||
| XM_005562561.2 | XP_005562618.1 | 3 - 4 | 293 | 97 | ||
| XM_015454430.1 | XP_015309916.1 | 5 - 6 | 651 | 97 | ||
| XM_015454431.1 | XP_015309917.1 | 3 - 4 | 470 | 97 | ||
| XM_015454432.1 | XP_015309918.1 | 3 - 4 | 473 | 97 | ||
| XM_015454433.1 | XP_015309919.1 | 5 - 6 | 655 | 97 | ||
| XM_015454434.1 | XP_015309920.1 | 5 - 6 | 657 | 97 | ||
Set Membership: |
|