Search
Search

C5H4orf47| Gene Symbol: C5H4orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556434.2 | XP_005556491.1 | 4 - 5 | 856 | 72 | |
| XM_005556436.2 | XP_005556493.1 | 4 - 5 | 859 | 72 | ||
Set Membership: |
|