Search
Search

LOC102126012| Gene Symbol: LOC102126012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577731.2 | XP_005577788.1 | 5 - 6 | 787 | 82 | |
| XM_005577733.2 | XP_005577790.1 | 5 - 6 | 765 | 82 | ||
Set Membership: |
|