Search
Search

ZNF215| Gene Symbol: ZNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578765.2 | XP_005578822.1 | 4 - 5 | 1301 | 75 | |
| XM_015435322.1 | XP_015290808.1 | 4 - 5 | 1301 | 75 | ||
Set Membership: |
|