Search
Search

CATSPERD| Gene Symbol: CATSPERD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587651.1 | XP_005587708.1 | 4 - 5 | 374 | 76 | |
| XM_005587652.2 | XP_005587709.1 | 4 - 5 | 352 | 76 | ||
| XM_005587653.2 | XP_005587710.1 | 4 - 5 | 352 | 76 | ||
| XM_005587654.2 | XP_005587711.1 | 4 - 5 | 352 | 76 | ||
| XM_005587655.2 | XP_005587712.1 | 4 - 5 | 352 | 76 | ||
| XM_015440278.1 | XP_015295764.1 | 3 - 4 | 283 | 76 | ||
| XM_015440279.1 | XP_015295765.1 | 3 - 4 | 255 | 76 | ||
| XM_015440280.1 | XP_015295766.1 | 4 - 5 | 225 | 76 | ||
Set Membership: |
|