Search
Search

TSPAN13| Gene Symbol: TSPAN13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319629.1 | NP_001306558.1 | 3 - 4 | 535 | 65 | |
| XM_005550051.2 | XP_005550108.2 | 3 - 4 | 766 | 65 | ||
| GenBank mRNA | AB174169.1 | - | 3 - 4 | 535 | 65 | |
Set Membership: |
|