Search
Search

WDR90| Gene Symbol: WDR90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442313.1 | XP_015297799.1 | 33 - 34 | 4793 | 87 | |
| XM_015442314.1 | XP_015297800.1 | 33 - 34 | 4793 | 87 | ||
| XM_015442315.1 | XP_015297801.1 | 34 - 35 | 4434 | 87 | ||
Set Membership: |
|