Search
Search

C15H9orf43| Gene Symbol: C15H9orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319373.1 | NP_001306302.1 | 7 - 8 | 1024 | 70 | |
| XM_005581007.2 | XP_005581064.1 | 7 - 8 | 931 | 70 | ||
| XM_005581009.2 | XP_005581066.1 | 8 - 9 | 995 | 70 | ||
| XM_005581010.2 | XP_005581067.1 | 7 - 8 | 927 | 70 | ||
| XM_005581011.2 | XP_005581068.1 | 7 - 8 | 736 | 70 | ||
| GenBank mRNA | AB070149.1 | - | 7 - 8 | 1025 | 70 | |
| AB168868.1 | - | 7 - 8 | 1024 | 70 | ||
Set Membership: |
|