Search
Search

LOC102122444| Gene Symbol: LOC102122444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319473.1 | NP_001306402.1 | 6 - 7 | 1061 | 105 | |
| XM_005539971.2 | XP_005540028.1 | 6 - 7 | 6624 | 105 | ||
| GenBank mRNA | KC734487.1 | - | 6 - 7 | 1061 | 105 | |
Set Membership: |
|