Search
Search

SCEL| Gene Symbol: SCEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586042.2 | XP_005586099.1 | 32 - 33 | 2222 | 116 | |
| XM_005586043.2 | XP_005586100.1 | 32 - 33 | 2218 | 116 | ||
| XM_005586044.2 | XP_005586101.1 | 32 - 33 | 2219 | 116 | ||
| XM_005586046.2 | XP_005586103.1 | 31 - 32 | 2162 | 116 | ||
| XM_005586047.2 | XP_005586104.1 | 31 - 32 | 2152 | 116 | ||
| XM_005586048.2 | XP_005586105.1 | 31 - 32 | 2131 | 116 | ||
| XM_005586049.2 | XP_005586106.1 | 31 - 32 | 2127 | 116 | ||
| XM_015439389.1 | XP_015294875.1 | 31 - 32 | 2162 | 116 | ||
| XM_015439390.1 | XP_015294876.1 | 30 - 31 | 2102 | 116 | ||
| XM_015439391.1 | XP_015294877.1 | 30 - 31 | 2067 | 116 | ||
| GenBank mRNA | AB179083.1 | - | 20 - 21 | 1411 | 116 | |
Set Membership: |
|