Search
Search

DIRAS2| Gene Symbol: DIRAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582222.2 | XP_005582279.1 | 2 | 2225 | 119 | |
| XM_015436571.1 | XP_015292057.1 | 2 | 2872 | 119 | ||
| GenBank mRNA | AB047874.1 | - | 1 | 518 | 119 | |
| AB062937.1 | - | 2 | 2238 | 119 | ||
| AB169837.1 | - | 977 | 119 | |||
Set Membership: |
|