Search
Search

CAPN14| Gene Symbol: CAPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576185.2 | XP_005576242.1 | 8 - 9 | 1230 | 74 | |
| XM_005576187.2 | XP_005576244.1 | 8 - 9 | 1230 | 74 | ||
| XM_005576188.2 | XP_005576245.1 | 9 - 10 | 1095 | 74 | ||
| XM_015433829.1 | XP_015289315.1 | 8 - 9 | 3187 | 74 | ||
Set Membership: |
|