Search
Search

ADCY7| Gene Symbol: ADCY7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591893.2 | XP_005591950.1 | 13 - 14 | 2106 | 103 | |
| XM_015443004.1 | XP_015298490.1 | 13 - 14 | 2022 | 103 | ||
| XM_015443005.1 | XP_015298491.1 | 13 - 14 | 2268 | 103 | ||
| XM_015443006.1 | XP_015298492.1 | 13 - 14 | 2107 | 103 | ||
| XM_015443007.1 | XP_015298493.1 | 13 - 14 | 2095 | 103 | ||
| XM_015443008.1 | XP_015298494.1 | 13 - 14 | 2308 | 103 | ||
Set Membership: |
|