Search
Search

MMRN1| Gene Symbol: MMRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555425.2 | XP_005555482.1 | 4 - 5 | 1155 | 67 | |
| XM_015450653.1 | XP_015306139.1 | 4 - 5 | 1148 | 67 | ||
Set Membership: |
|