Search
Search

CPAMD8| Gene Symbol: CPAMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015440499.1 | XP_015295985.1 | 18 - 19 | 3061 | 78 | |
| XM_015440500.1 | XP_015295986.1 | 18 - 19 | 3061 | 78 | ||
Set Membership: |
|