Search
Search

BUD13| Gene Symbol: BUD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579721.2 | XP_005579778.1 | 4 - 5 | 1088 | 77 | |
| XM_015435747.1 | XP_015291233.1 | 4 - 5 | 1037 | 77 | ||
| XM_015435748.1 | XP_015291234.1 | 5 - 6 | 1089 | 77 | ||
| XM_015435749.1 | XP_015291235.1 | 5 - 6 | 1089 | 77 | ||
| XM_015435750.1 | XP_015291236.1 | 4 - 5 | 1012 | 77 | ||