Search
Search

GGACT| Gene Symbol: GGACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439477.1 | XP_015294963.1 | 2 | 3319 | 121 | |
| XM_015439478.1 | XP_015294964.1 | 4 | 810 | 121 | ||
| XM_015439479.1 | XP_015294965.1 | 2 | 559 | 121 | ||
| XM_015439480.1 | XP_015294966.1 | 4 | 1068 | 121 | ||
| XM_015439481.1 | XP_015294967.1 | 5 | 887 | 121 | ||
| XM_015439482.1 | XP_015294968.1 | 3 | 548 | 121 | ||
Set Membership: |
|