Search
Search
| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007423.4 | NP_031449.3 | 4 - 5 | 533 | 73 | |
| GenBank mRNA | AK010934.1 | - | 4 - 5 | 509 | 73 | |
| AK075972.1 | - | 4 - 5 | 513 | 73 | ||
| AK076053.1 | - | 4 - 5 | 509 | 73 | ||
| AK076197.1 | - | 4 - 5 | 511 | 73 | ||
| AK145975.1 | - | 4 - 5 | 509 | 73 | ||
| AK146513.1 | - | 4 - 5 | 511 | 73 | ||
| AK146524.1 | - | 4 - 5 | 511 | 73 | ||
| AK167549.1 | - | 4 - 5 | 509 | 73 | ||
| AK168426.1 | - | 4 - 5 | 511 | 73 | ||
| AK168574.1 | - | 4 - 5 | 511 | 73 | ||
| AK168576.1 | - | 4 - 5 | 509 | 73 | ||
| AK168751.1 | - | 4 - 5 | 511 | 73 | ||
| AK168818.1 | - | 3 - 4 | 387 | 73 | ||
| AK168883.1 | - | 4 - 5 | 509 | 73 | ||
| AK169046.1 | - | 4 - 5 | 511 | 73 | ||
| AK169054.1 | - | 4 - 5 | 511 | 73 | ||
| AK169147.1 | - | 4 - 5 | 509 | 73 | ||
| BC066206.1 | - | 4 - 5 | 496 | 30469872 | 73 | |
| V00743.1 | - | 4 - 5 | 507 | 73 | ||
Set Membership: |
|