Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432265_g1
          See other DDR1 GE Assays ›
          Gene Symbol:
          Ddr1
          Gene Name:
          discoidin domain receptor family, member 1
          Gene Aliases:
          6030432F18, AI323681, CD167a, Cak, Nep, PTK3A
          Chromosome Location:
          Chr.17: 35681567 - 35704629 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432265_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198831.1 NP_001185760.1 15 - 16 2288 112
          NM_001198833.1 NP_001185762.1 15 - 16 2407 112
          NM_007584.2 NP_031610.1 15 - 16 2463 112
          NM_172962.1 NP_766550.1 14 - 15 2352 112
          GenBank mRNA AF026259.1 - 15 - 16 2305 112
          AK031442.1 - 14 - 15 2255 112
          AK138698.1 - 15 - 16 2276 112
          AK143088.1 - 14 - 15 2245 112
          BC006836.1 - 13 - 14 2155 112
          BC037108.1 - 16 - 17 2542 112
          L57509.1 - 15 - 16 2311 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001976.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddr1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          regulation of cell-matrix adhesion
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          female pregnancy
          embryo implantation
          lactation
          negative regulation of cell proliferation
          regulation of extracellular matrix disassembly
          smooth muscle cell migration
          phosphorylation
          peptidyl-tyrosine phosphorylation
          collagen-activated tyrosine kinase receptor signaling pathway
          peptidyl-tyrosine autophosphorylation
          ear development
          wound healing, spreading of cells
          protein autophosphorylation
          branching involved in mammary gland duct morphogenesis
          mammary gland alveolus development
          smooth muscle cell-matrix adhesion
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          collagen binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein tyrosine kinase collagen receptor activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_001198831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_001198831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_001198833.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_001198833.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_007584.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_007584.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_172962.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 NM_172962.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018806.mm.1 TC1700001976.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline