Search
Search

Eef2k| Gene Symbol: Eef2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267710.1 | NP_001254639.1 | 7 - 8 | 1025 | 74 | |
| NM_001267711.1 | NP_001254640.1 | 7 - 8 | 1020 | 74 | ||
| NM_007908.4 | NP_031934.1 | 7 - 8 | 1109 | 74 | ||
| GenBank mRNA | AK028524.1 | - | 7 - 8 | 1086 | 74 | |
| AK080915.1 | - | 7 - 8 | 1026 | 74 | ||
| AK135891.1 | - | 5 - 6 | 451 | 74 | ||
| AK136015.1 | - | 7 - 8 | 1021 | 74 | ||
| AK177226.1 | - | 2 - 3 | 243 | 75 | ||
| AK188679.1 | - | 2 - 3 | 243 | 74 | ||
| AK208051.1 | - | 2 - 3 | 257 | 74 | ||
| AK212025.1 | - | 2 - 3 | 244 | 74 | ||
| BC003433.1 | - | 7 - 8 | 1057 | 2646503 | 74 | |
| BC055361.1 | - | 7 - 8 | 1020 | 30046537 | 74 | |
| U93848.1 | - | 6 - 7 | 763 | 74 | ||
Set Membership: |
|