Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433881_g1
          See other GUCY2E GE Assays ›
          Gene Symbol:
          Gucy2e
          Gene Name:
          guanylate cyclase 2e
          Gene Aliases:
          GC-E, GC1, ROS-GC1
          Chromosome Location:
          Chr.11: 69218117 - 69237358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433881_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gucy2e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008192.3 NP_032218.2 16 - 17 3138 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003034.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gucy2e

          Panther Classification:

          Molecular Function -

          guanylate cyclase

          Gene Ontology Categories:

          Function(s) Process(es)

          cGMP biosynthetic process
          protein phosphorylation
          visual perception
          cyclic nucleotide biosynthetic process
          intracellular signal transduction
          response to stimulus
          nucleotide binding
          guanylate cyclase activity
          protein kinase activity
          ATP binding
          GTP binding
          transferase activity, transferring phosphorus-containing groups
          lyase activity
          phosphorus-oxygen lyase activity
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100027549.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027549.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027549.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027553.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027553.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027553.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027555.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027555.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027555.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027552.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027552.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027552.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027556.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027556.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027556.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027554.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027554.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027554.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027550.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027550.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027550.mm.1 TC1100003034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027551.mm.1 NM_008192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027551.mm.1 NM_008192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027551.mm.1 TC1100003034.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline