Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434268_m1
          See other IL2RB GE Assays ›
          Gene Symbol:
          Il2rb
          Gene Name:
          interleukin 2 receptor, beta chain
          Gene Aliases:
          CD122, IL-15Rbeta, IL15Rbeta, Il-2/15Rbeta, Il-2Rbeta, p70
          Chromosome Location:
          Chr.15: 78479256 - 78511627 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434268_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il2rb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008368.4 NP_032394.1 5 - 6 544 64
          GenBank mRNA AK017288.1 - 5 - 6 544 64
          AK035289.1 - 5 - 6 497 64
          AK157572.1 - 5 - 6 545 64
          AK169481.1 - 5 - 6 525 64
          BC026869.1 - 5 - 6 517 4015306 64
          BC099616.1 - 5 - 6 30024496 64
          M28052.1 - 5 - 6 501 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001833.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il2rb

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          natural killer cell activation
          interleukin-2-mediated signaling pathway
          negative regulation of apoptotic process
          cytokine receptor activity
          interleukin-2 receptor activity
          interleukin-2 binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 BC026869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 BQ033566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 AK169481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 AI047841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 AK017288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 BC026869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 BQ033566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 AK169481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 AI047841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286496 AK017288.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172229 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 BC026869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 BQ033566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 AK169481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 AI047841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 AK017288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 BC026869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 BQ033566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 AK169481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 AI047841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286496 AK017288.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 BC026869.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 BQ033566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 AK169481.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 AI047841.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 AK017288.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 BC026869.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 BQ033566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 AK169481.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 AI047841.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286496 AK017288.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30360431 - -
          Whole Mouse Genome G4122A A_51_P286496 BC026869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 BQ033566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 AK169481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 AI047841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 AK017288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 BC026869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 BQ033566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 AK169481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 AI047841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286496 AK017288.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11946941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11989845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12298074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12298072 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11989848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30360244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12286357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12299337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12153711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12345709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12299336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12209907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12299334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12299329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12211300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12153699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12284289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30360421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12211306 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430347 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430351 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430350 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430349 - -
          Affymetrix HT MG-430 PM Array Plate 1448759_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417546_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430344 - -
          Mouse Genome 430 2.0 Array 1417546_at - -
          Mouse Genome 430 2.0 Array 1417546_PM_at - -
          Mouse Genome 430 2.0 Array 1459870_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448759_at - -
          Mouse Genome 430 2.0 Array 1459870_x_at - -
          Mouse Genome 430 2.0 Array 1448759_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16185_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16185_at - -
          Affymetrix Mouse Clariom S Array PSR1500014344.mm.1 NM_008368.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014344.mm.1 NM_008368.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014344.mm.1 TC1500001833.mm.1 MTA TC
          Mouse Expression Set 430 1417546_at - -
          Mouse Expression Set 430 1448759_PM_at - -
          Mouse Expression Set 430 1417546_PM_at - -
          Mouse Expression Set 430 1448759_at - -
          Murine Genome U74v2 Set 100764_PM_at - -
          Murine Genome U74v2 Set 100764_at - -
          Mouse Genome 430A 2.0 Array 1417546_PM_at - -
          Mouse Genome 430A 2.0 Array 1448759_at - -
          Mouse Genome 430A 2.0 Array 1448759_PM_at - -
          Mouse Genome 430A 2.0 Array 1417546_at - -
          Affymetrix HT Mouse Genome 430A Array 1417546_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448759_at - -
          Affymetrix HT Mouse Genome 430A Array 1417546_at - -
          Affymetrix HT Mouse Genome 430A Array 1448759_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4730072 - -
          Illumina mouse-6 v1.0 expression beadchip 4730072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246541 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246541 - -
          mouse-6 v1.1 (Illumina) scl46990.11.1_92-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46990.11.1_92-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246541 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46990.11.1_92-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline