Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434669_m1
          See other SERPINA3C GE Assays ›
          Gene Symbol:
          Serpina3c
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A, member 3C
          Gene Aliases:
          1A1, Kalbp, Klkbp, spi2
          Chromosome Location:
          Chr.12: 104146907 - 104153933 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434669_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina3c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008458.2 NP_032484.1 1 - 2 98 64
          GenBank mRNA BC116724.1 - 1 - 2 96 40106394 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002289.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC116724.1,NM_008458.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina3c

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          response to cytokine
          response to peptide hormone
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P157112 BC116724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P157112 NM_008458.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P157112 BC116724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P157112 NM_008458.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105156 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158727 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P157112 BC116724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P157112 NM_008458.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P157112 BC116724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P157112 NM_008458.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158727 - -
          Whole Mouse Genome G4122A A_51_P157112 BC116724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157112 NM_008458.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157112 BC116724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157112 NM_008458.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125978 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158727 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10371571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10371581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10371579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10371579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10371570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10292417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402439 - -
          Affymetrix HT MG-430 PM Array Plate 1421564_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10402435 - -
          Mouse Genome 430 2.0 Array 1421564_at - -
          Mouse Genome 430 2.0 Array 1421564_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16625_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16625_at - -
          Affymetrix Mouse Clariom S Array PSR1200015955.mm.1 NM_008458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015955.mm.1 NM_008458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015955.mm.1 TC1200002289.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200015957.mm.1 NM_008458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015957.mm.1 NM_008458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015957.mm.1 TC1200002289.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200015958.mm.1 NM_008458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015958.mm.1 NM_008458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015958.mm.1 TC1200002289.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200015959.mm.1 NM_008458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015959.mm.1 NM_008458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015959.mm.1 TC1200002289.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200015956.mm.1 NM_008458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015956.mm.1 NM_008458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015956.mm.1 TC1200002289.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200015960.mm.1 NM_008458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015960.mm.1 NM_008458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015960.mm.1 TC1200002289.mm.1 MTA TC
          Mouse Expression Set 430 1421564_PM_at - -
          Mouse Expression Set 430 1421564_at - -
          Murine Genome U74v2 Set 102707_PM_f_at - -
          Murine Genome U74v2 Set 102706_i_at - -
          Murine Genome U74v2 Set 102706_PM_i_at - -
          Murine Genome U74v2 Set 102707_f_at - -
          Mouse Genome 430A 2.0 Array 1421564_PM_at - -
          Mouse Genome 430A 2.0 Array 1421564_at - -
          Affymetrix HT Mouse Genome 430A Array 1421564_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421564_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 643186 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2730054 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2730054 - -
          mouse-6 v1.1 (Illumina) scl42101.2_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42101.2_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2730054 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2730054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline