Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434764_m1
          See other LPL GE Assays ›
          Gene Symbol:
          Lpl
          Gene Name:
          lipoprotein lipase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 68880555 - 68906932 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434764_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lpl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008509.2 NP_032535.2 2 - 3 447 61
          GenBank mRNA AK002645.1 - 2 - 3 373 61
          AK045064.1 - 2 - 3 336 61
          AK079446.1 - 2 - 3 336 61
          AK086023.1 - 2 - 3 268 61
          AK150328.1 - 2 - 3 448 61
          AK150355.1 - 2 - 3 448 61
          AK150375.1 - 2 - 3 446 61
          AK150457.1 - 2 - 3 343 61
          AK150488.1 - 2 - 3 448 61
          AK150505.1 - 2 - 3 448 61
          AK150532.1 - 2 - 3 448 61
          AK150577.1 - 2 - 3 448 61
          AK150585.1 - 2 - 3 448 61
          AK150593.1 - 2 - 3 448 61
          AK150646.1 - 2 - 3 445 61
          AK150672.1 - 2 - 3 366 61
          AK150691.1 - 2 - 3 448 61
          AK150984.1 - 2 - 3 448 61
          AK150990.1 - 2 - 3 448 61
          AK151006.1 - 2 - 3 448 61
          AK151038.1 - 2 - 3 346 61
          AK151049.1 - 2 - 3 448 61
          AK151093.1 - 2 - 3 448 61
          AK151105.1 - 2 - 3 448 61
          AK151112.1 - 2 - 3 449 61
          AK151195.1 - 2 - 3 346 61
          AK151216.1 - 2 - 3 449 61
          AK151369.1 - 2 - 3 366 61
          AK151434.1 - 2 - 3 366 61
          AK151521.1 - 2 - 3 366 61
          AK151540.1 - 2 - 3 336 61
          AK151563.1 - 2 - 3 448 61
          AK151630.1 - 2 - 3 346 61
          AK151679.1 - 2 - 3 448 61
          AK151698.1 - 2 - 3 448 61
          AK151727.1 - 2 - 3 448 61
          AK151772.1 - 2 - 3 448 61
          AK151774.1 - 2 - 3 346 61
          AK151801.1 - 2 - 3 336 61
          AK151828.1 - 2 - 3 336 61
          AK151866.1 - 2 - 3 448 61
          AK151870.1 - 2 - 3 448 61
          AK151872.1 - 2 - 3 448 61
          AK151893.1 - 2 - 3 336 61
          AK152014.1 - 2 - 3 448 61
          AK152035.1 - 2 - 3 448 61
          AK152049.1 - 2 - 3 448 61
          AK152053.1 - 2 - 3 336 61
          AK152079.1 - 2 - 3 448 61
          AK152238.1 - 2 - 3 448 61
          AK152251.1 - 2 - 3 445 61
          AK152350.1 - 2 - 3 448 61
          AK152390.1 - 2 - 3 287 61
          AK152400.1 - 2 - 3 346 61
          AK152481.1 - 2 - 3 449 61
          AK152657.1 - 2 - 3 448 61
          AK152706.1 - 2 - 3 449 61
          AK152733.1 - 2 - 3 346 61
          AK152775.1 - 2 - 3 448 61
          AK152869.1 - 2 - 3 448 61
          AK152956.1 - 2 - 3 448 61
          AK153006.1 - 2 - 3 448 61
          AK153148.1 - 2 - 3 448 61
          AK153242.1 - 2 - 3 449 61
          AK153425.1 - 2 - 3 448 61
          AK153428.1 - 2 - 3 448 61
          AK159268.1 - 2 - 3 448 61
          AK170486.1 - 2 - 3 448 61
          BC003305.1 - 2 - 3 436 3584788 61
          BC158040.1 - 2 - 3 281 9088156 61
          CT010344.1 - 2 - 3 249 61
          J03302.1 - 2 - 3 225 61
          M65258.1 - 2 - 3 409 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000707.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lpl

          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid biosynthetic process
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          positive regulation of sequestering of triglyceride
          lipid catabolic process
          triglyceride biosynthetic process
          triglyceride catabolic process
          very-low-density lipoprotein particle remodeling
          triglyceride homeostasis
          lipoprotein lipase activity
          triglyceride lipase activity
          receptor binding
          protein binding
          heparin binding
          hydrolase activity
          triglyceride binding
          apolipoprotein binding
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800004738.mm.1 NM_008509.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004738.mm.1 NM_008509.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004738.mm.1 TC0800000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004742.mm.1 NM_008509.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004742.mm.1 NM_008509.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004742.mm.1 TC0800000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004739.mm.1 NM_008509.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004739.mm.1 NM_008509.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004739.mm.1 TC0800000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004747.mm.1 NM_008509.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004747.mm.1 NM_008509.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004747.mm.1 TC0800000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004737.mm.1 NM_008509.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004737.mm.1 NM_008509.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004737.mm.1 TC0800000707.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline