Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435217_m1
          See other NOS3 GE Assays ›
          Gene Symbol:
          Nos3
          Gene Name:
          nitric oxide synthase 3, endothelial cell
          Gene Aliases:
          2310065A03Rik, Nos-3, eNOS, ecNOS
          Chromosome Location:
          Chr.5: 24364819 - 24384474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435217_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nos3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008713.4 NP_032739.3 22 - 23 3002 71
          GenBank mRNA AK077896.1 - 22 - 23 3004 71
          AK143221.1 - 21 - 22 2775 71
          AK158106.1 - 22 - 23 3025 71
          BC052636.1 - 22 - 23 2989 6530765 71
          U53142.1 - 22 - 23 2990 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000212.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nos3

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovulation from ovarian follicle
          in utero embryonic development
          blood vessel remodeling
          regulation of sodium ion transport
          regulation of the force of heart contraction by chemical signal
          regulation of systemic arterial blood pressure by endothelin
          arginine catabolic process
          nitric oxide biosynthetic process
          signal transduction
          nitric oxide mediated signal transduction
          negative regulation of cell proliferation
          negative regulation of muscle hyperplasia
          smooth muscle hyperplasia
          lung development
          positive regulation of guanylate cyclase activity
          lipopolysaccharide-mediated signaling pathway
          response to estradiol
          positive regulation of apoptotic process
          negative regulation of potassium ion transport
          endothelial cell migration
          positive regulation of angiogenesis
          negative regulation of blood pressure
          positive regulation of vasodilation
          negative regulation of smooth muscle cell proliferation
          regulation of blood vessel size
          negative regulation of hydrolase activity
          negative regulation of calcium ion transport
          oxidation-reduction process
          cellular response to mechanical stimulus
          cellular response to transforming growth factor beta stimulus
          actin binding
          actin monomer binding
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          iron ion binding
          calmodulin binding
          beta-catenin binding
          FMN binding
          oxidoreductase activity
          heme binding
          tetrahydrobiopterin binding
          arginine binding
          cadherin binding
          metal ion binding
          flavin adenine dinucleotide binding
          NADP binding
          nitric-oxide synthase binding
          Hsp90 protein binding
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500001542.mm.1 TC0500000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001542.mm.1 NM_008713.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001542.mm.1 NM_008713.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline