Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435362_m1
          See other SLC11A2 GE Assays ›
          Gene Symbol:
          Slc11a2
          Gene Name:
          solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2
          Gene Aliases:
          DCT1, DMT1, Nramp2, mk, van
          Chromosome Location:
          Chr.15: 100387898 - 100424209 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435362_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc11a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146161.1 NP_001139633.1 14 - 15 1560 88
          NM_008732.2 NP_032758.2 14 - 15 1560 88
          GenBank mRNA AF029758.1 - 14 - 15 1495 88
          AK049856.1 - 14 - 15 1508 88
          AK083478.1 - 14 - 15 1506 88
          AK148276.1 - 14 - 15 1508 88
          BC019137.1 - 14 - 15 1534 5040976 88
          L33415.1 - 14 - 15 1488 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002238.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc11a2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transition metal ion transport
          porphyrin-containing compound metabolic process
          porphyrin-containing compound biosynthetic process
          heme biosynthetic process
          transport
          ion transport
          cobalt ion transport
          copper ion transport
          iron ion transport
          manganese ion transport
          cellular copper ion homeostasis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          learning or memory
          response to manganese ion
          nickel cation transport
          vanadium ion transport
          ferrous iron transport
          lead ion transport
          proton transport
          cellular response to oxidative stress
          iron ion transmembrane transport
          copper ion transmembrane transport
          nickel cation transmembrane transport
          dendrite morphogenesis
          erythrocyte development
          iron ion homeostasis
          multicellular organismal iron ion homeostasis
          cadmium ion transmembrane transport
          ferrous iron import
          manganese ion transmembrane transport
          zinc II ion transmembrane transport
          cation transmembrane transport
          hydrogen ion transmembrane transport
          copper ion import into cell
          ferrous iron transmembrane transport
          transporter activity
          copper ion transmembrane transporter activity
          iron ion transmembrane transporter activity
          manganese ion transmembrane transporter activity
          iron ion binding
          copper ion binding
          zinc ion binding
          hydrogen ion transmembrane transporter activity
          cadmium ion transmembrane transporter activity
          cobalt ion transmembrane transporter activity
          ferrous iron transmembrane transporter activity
          lead ion transmembrane transporter activity
          nickel cation transmembrane transporter activity
          vanadium ion transmembrane transporter activity
          solute:proton symporter activity
          ferrous iron uptake transmembrane transporter activity
          nickel cation binding
          inorganic cation transmembrane transporter activity
          manganese ion binding
          cadmium ion binding
          transition metal ion transmembrane transporter activity
          cobalt ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500018313.mm.1 NM_001146161.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018313.mm.1 NM_001146161.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018313.mm.1 NM_008732.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018313.mm.1 NM_008732.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018313.mm.1 TC1500002238.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018314.mm.1 NM_001146161.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018314.mm.1 NM_001146161.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018314.mm.1 NM_008732.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018314.mm.1 NM_008732.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018314.mm.1 TC1500002238.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline