Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436527_m1
          See other SHH GE Assays ›
          Gene Symbol:
          Shh
          Gene Name:
          sonic hedgehog
          Gene Aliases:
          9530036O11Rik, Dsh, Hhg1, Hx, Hxl3, M100081
          Chromosome Location:
          Chr.5: 28456840 - 28467101 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436527_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009170.3 NP_033196.1 1 - 2 637 105
          GenBank mRNA AK077688.1 - 1 - 2 638 105
          BC063087.1 - 1 - 2 624 6516263 105
          EU664592.1 - 1 - 2 183 105
          X76290.1 - 1 - 2 299 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002136.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Mouse Androgen Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shh

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          patterning of blood vessels
          vasculogenesis
          metanephros development
          branching involved in ureteric bud morphogenesis
          cell fate specification
          neural crest cell migration
          kidney development
          neural tube formation
          hair follicle development
          vasculature development
          heart looping
          positive regulation of neuroblast proliferation
          positive regulation of mesenchymal cell proliferation
          osteoblast development
          lymphoid progenitor cell differentiation
          determination of left/right asymmetry in lateral mesoderm
          regulation of transcription, DNA-templated
          proteolysis
          endocytosis
          apoptotic process
          cell communication
          signal transduction
          smoothened signaling pathway
          positive regulation of hh target transcription factor activity
          cell-cell signaling
          multicellular organism development
          determination of left/right symmetry
          pattern specification process
          ectoderm development
          neuroblast proliferation
          axon guidance
          central nervous system development
          hindgut morphogenesis
          digestive tract mesoderm development
          heart development
          blood coagulation
          androgen metabolic process
          cell proliferation
          positive regulation of cell proliferation
          embryo development
          polarity specification of anterior/posterior axis
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          mesenchymal cell proliferation
          regulation of gene expression
          positive regulation of gene expression
          negative regulation of gene expression
          oligodendrocyte development
          striated muscle tissue development
          positive regulation of skeletal muscle cell proliferation
          myotube differentiation
          intein-mediated protein splicing
          spinal cord dorsal/ventral patterning
          ventral spinal cord interneuron specification
          spinal cord motor neuron differentiation
          thalamus development
          dorsal/ventral neural tube patterning
          cerebellar granule cell precursor proliferation
          smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
          positive regulation of cerebellar granule cell precursor proliferation
          telencephalon regionalization
          establishment of cell polarity
          regulation of proteolysis
          positive regulation of Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          respiratory tube development
          lung development
          embryonic limb morphogenesis
          negative regulation of cell migration
          male genitalia development
          prostate gland development
          thyroid gland development
          forebrain development
          midbrain development
          hindbrain development
          pancreas development
          hair follicle morphogenesis
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          T cell differentiation in thymus
          positive regulation of T cell differentiation in thymus
          positive regulation of immature T cell proliferation in thymus
          negative regulation of transcription elongation from RNA polymerase II promoter
          protein localization to nucleus
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          regulation of cell proliferation
          negative regulation of T cell proliferation
          negative regulation of protein catabolic process
          positive regulation of protein import into nucleus
          odontogenesis of dentin-containing tooth
          odontogenesis
          regulation of odontogenesis
          embryonic digit morphogenesis
          camera-type eye development
          negative regulation of apoptotic process
          CD4-positive or CD8-positive, alpha-beta T cell lineage commitment
          tongue development
          tongue morphogenesis
          skin development
          positive thymic T cell selection
          negative thymic T cell selection
          intermediate filament organization
          cell fate commitment
          myoblast differentiation
          response to ethanol
          negative regulation of cell differentiation
          positive regulation of cell differentiation
          positive regulation of neuron differentiation
          positive regulation of smoothened signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of photoreceptor cell differentiation
          positive regulation of alpha-beta T cell differentiation
          negative regulation of alpha-beta T cell differentiation
          cell development
          thymus development
          digestive tract morphogenesis
          embryonic digestive tract morphogenesis
          embryonic organ development
          developmental growth
          embryonic morphogenesis
          embryonic foregut morphogenesis
          positive regulation of skeletal muscle tissue development
          organ formation
          anatomical structure formation involved in morphogenesis
          neuron fate commitment
          response to axon injury
          embryonic skeletal system development
          oligodendrocyte differentiation
          positive regulation of oligodendrocyte differentiation
          branching morphogenesis of an epithelial tube
          male genitalia morphogenesis
          inner ear development
          anatomical structure development
          formation of anatomical boundary
          stem cell development
          striated muscle cell differentiation
          positive regulation of striated muscle cell differentiation
          positive regulation of cell division
          Bergmann glial cell differentiation
          palate development
          canonical Wnt signaling pathway
          limb development
          limb bud formation
          positive regulation of penile erection
          lung morphogenesis
          lung epithelium development
          trachea development
          trachea morphogenesis
          epithelial tube branching involved in lung morphogenesis
          branching involved in prostate gland morphogenesis
          branching involved in salivary gland morphogenesis
          bud outgrowth involved in lung branching
          right lung development
          left lung development
          lung lobe morphogenesis
          lung-associated mesenchyme development
          prostatic bud formation
          primary prostatic bud elongation
          prostate epithelial cord elongation
          salivary gland cavitation
          epithelial cell proliferation involved in salivary gland morphogenesis
          epithelial-mesenchymal cell signaling
          regulation of prostatic bud formation
          epithelial-mesenchymal signaling involved in prostate gland development
          regulation of epithelial cell proliferation involved in prostate gland development
          positive regulation of epithelial cell proliferation involved in prostate gland development
          regulation of mesenchymal cell proliferation involved in prostate gland development
          mesenchymal smoothened signaling pathway involved in prostate gland development
          artery development
          mesenchymal cell proliferation involved in lung development
          vasculogenesis involved in coronary vascular morphogenesis
          somite development
          positive regulation of sclerotome development
          fungiform papilla development
          fungiform papilla morphogenesis
          fungiform papilla formation
          positive regulation of oligodendrocyte progenitor proliferation
          cellular response to lithium ion
          metanephric mesenchymal cell proliferation involved in metanephros development
          multicellular structure septum development
          negative regulation of canonical Wnt signaling pathway
          negative regulation of cholesterol efflux
          apoptotic signaling pathway
          regulation of protein localization to nucleus
          negative regulation of ureter smooth muscle cell differentiation
          positive regulation of ureter smooth muscle cell differentiation
          negative regulation of kidney smooth muscle cell differentiation
          positive regulation of kidney smooth muscle cell differentiation
          positive regulation of mesenchymal cell proliferation involved in ureter development
          negative regulation of mesenchymal cell apoptotic process
          glycoprotein binding
          patched binding
          calcium ion binding
          protein binding
          glycosaminoglycan binding
          peptidase activity
          zinc ion binding
          hydrolase activity
          laminin-1 binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500019423.mm.1 NM_009170.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019423.mm.1 NM_009170.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019423.mm.1 TC0500002136.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019424.mm.1 NM_009170.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019424.mm.1 NM_009170.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019424.mm.1 TC0500002136.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline