Search
Search

Tgfbr2| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009371.3 | NP_033397.3 | 2 - 3 | 491 | 73 | |
| GenBank mRNA | AK043619.1 | - | 2 - 3 | 575 | 73 | |
| AK053793.1 | - | 2 - 3 | 492 | 73 | ||
| AK159327.1 | - | 2 - 3 | 471 | 73 | ||
| BC052629.1 | - | 393 | 73 | |||
Set Membership: |
|