Search
Search

Nr1h2| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285518.1 | NP_001272447.1 | 2 - 3 | 283 | 94 | |
| NM_009473.3 | NP_033499.1 | 2 - 3 | 287 | 94 | ||
| GenBank mRNA | AK134489.1 | - | 2 - 3 | 242 | 94 | |
| BQ573162.1 | - | 2 - 3 | 257 | 94 | ||
| BY197597.1 | - | 2 - 3 | 286 | 94 | ||
| U09419.1 | - | 1 - 2 | 105 | 94 | ||
Set Membership: |
|