Search
Search

Acvrl1| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277255.1 | NP_001264184.1 | 10 - 11 | 1540 | 57 | |
| NM_001277257.1 | NP_001264186.1 | 9 - 10 | 1479 | 57 | ||
| NM_001277258.1 | NP_001264187.1 | 9 - 10 | 1448 | 57 | ||
| NM_001277259.1 | NP_001264188.1 | 10 - 11 | 1535 | 57 | ||
| NM_009612.3 | NP_033742.2 | 9 - 10 | 1517 | 57 | ||
| GenBank mRNA | AK134712.1 | - | 7 - 8 | 1274 | 57 | |
| AK160915.1 | - | 10 - 11 | 1508 | 57 | ||
| AK170870.1 | - | 7 - 8 | 1273 | 57 | ||
| BC015083.1 | - | 8 - 9 | 1398 | 3980151 | 57 | |
| L48015.1 | - | 9 - 10 | 1519 | 57 | ||
| Z31664.1 | - | 9 - 10 | 1485 | 57 | ||
Set Membership: |
|