Search
Search

Asgr1| Gene Symbol: Asgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291131.1 | NP_001278060.1 | 7 - 8 | 637 | 108 | |
| NM_001291132.1 | NP_001278061.1 | 6 - 7 | 660 | 108 | ||
| NM_009714.3 | NP_033844.1 | 7 - 8 | 747 | 108 | ||
| GenBank mRNA | AK132959.1 | - | 7 - 8 | 638 | 108 | |
| BC014779.1 | - | 7 - 8 | 653 | 108 | ||
| BC022106.1 | - | 7 - 8 | 709 | 5102469 | 108 | |
| U08372.1 | - | 6 - 7 | 797 | 108 | ||
| U09362.1 | - | 7 - 8 | 698 | 108 | ||
Set Membership: |
|