Search
Search

Atp2a2| Gene Symbol: Atp2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110140.3 | NP_001103610.1 | 18 - 19 | 3282 | 90 | |
| NM_009722.3 | NP_033852.1 | 18 - 19 | 3282 | 90 | ||
| NR_027838.1 | 18 - 19 | 3282 | 90 | |||
| GenBank mRNA | AJ131821.1 | - | 18 - 19 | 2952 | 90 | |
| AJ223584.1 | - | 18 - 19 | 2952 | 90 | ||
| AK017877.1 | - | 5 - 6 | 708 | 90 | ||
| AK181503.1 | - | 2 - 3 | 164 | 90 | ||
| AK212496.1 | - | 2 - 3 | 162 | 90 | ||
| AK220538.1 | - | 18 - 19 | 3231 | 90 | ||
| BC054531.1 | - | 18 - 19 | 2941 | 6406916 | 90 | |
| BC054748.1 | - | 18 - 19 | 2941 | 6406916 | 90 | |
Set Membership: |
|