Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437761_g1
          See other AVP GE Assays ›
          Gene Symbol:
          Avp
          Gene Name:
          arginine vasopressin
          Gene Aliases:
          Vp, Vsp
          Chromosome Location:
          Chr.2: 130580677 - 130582541 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437761_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Avp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009732.1 NP_033862.1 2 - 3 338 80
          GenBank mRNA AK138179.1 - 2 - 3 371 80
          BC051997.1 - 2 - 3 351 5683720 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004630.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Avp

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          maternal aggressive behavior
          positive regulation of systemic arterial blood pressure
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          negative regulation of female receptivity
          grooming behavior
          locomotory behavior
          positive regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of glutamate secretion
          positive regulation of cell growth
          positive regulation of cAMP biosynthetic process
          positive regulation of prostaglandin biosynthetic process
          positive regulation of cellular pH reduction
          positive regulation of peptidyl-serine phosphorylation
          social behavior
          regulation of renal sodium excretion
          vasoconstriction
          maternal behavior
          negative regulation of apoptotic process
          penile erection
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of vasoconstriction
          regulation of blood vessel size
          multicellular organismal water homeostasis
          negative regulation of transmission of nerve impulse
          ERK1 and ERK2 cascade
          protein kinase C signaling
          negative regulation of release of cytochrome c from mitochondria
          protein kinase activity
          signal transducer activity
          hormone activity
          neuropeptide hormone activity
          neurohypophyseal hormone activity
          V1A vasopressin receptor binding
          V1B vasopressin receptor binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630555 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630551 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136508 AK138179.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136508 BC051997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136508 NM_009732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136508 AK138179.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136508 BC051997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136508 NM_009732.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164168 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136508 AK138179.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136508 BC051997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136508 NM_009732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136508 AK138179.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136508 BC051997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136508 NM_009732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136508 AK138179.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136508 BC051997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136508 NM_009732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136508 AK138179.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136508 BC051997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136508 NM_009732.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630606 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164168 - -
          Whole Mouse Genome G4122A A_51_P136508 AK138179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136508 BC051997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136508 NM_009732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136508 AK138179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136508 BC051997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136508 NM_009732.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135730 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164168 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01630329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01630304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01630300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01630307 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01630327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01729553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630551 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01224764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01252950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01224766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01287729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01252954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01252952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01287732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01552171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01448450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10487686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487687 - -
          Affymetrix HT MG-430 PM Array Plate 1450794_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10487685 - -
          Mouse Genome 430 2.0 Array 1450794_PM_at - -
          Mouse Genome 430 2.0 Array 1450794_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11998_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11998_at - -
          Affymetrix Mouse Clariom S Array PSR0200039594.mm.1 TC0200004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039595.mm.1 NM_009732.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039595.mm.1 TC0200004630.mm.1 MTA TC
          Mouse Expression Set 430 1450794_at - -
          Mouse Expression Set 430 1450794_PM_at - -
          Murine Genome U74v2 Set 93381_PM_at - -
          Murine Genome U74v2 Set 93381_at - -
          Mouse Genome 430A 2.0 Array 1450794_PM_at - -
          Mouse Genome 430A 2.0 Array 1450794_at - -
          Affymetrix HT Mouse Genome 430A Array 1450794_at - -
          Affymetrix HT Mouse Genome 430A Array 1450794_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838715 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2100113 - -
          Illumina mouse-6 v1.0 expression beadchip 2100113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002100113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710928 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710928 - -
          mouse-6 v1.1 (Illumina) scl18662.3.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 2100113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18662.3.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2100113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710928 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710928 - -

          Back To Top
           

          Citations (1)

          An indispensable role of secretin in mediating the osmoregulatory functions of angiotensin II.
          Authors
          Lee VH, Lee LT, Chu JY, Lam IP, Siu FK, Vaudry H, Chow BK
          Journal
          Faseb J - Page No 5024 (Wed Dec 01 00:00:00 UTC 2010)
          Volume (Year)
          24 (2010)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline