Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437762_m1
          See other B2M GE Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          Ly-m11, beta2-m, beta2m
          Chromosome Location:
          Chr.2: 122147687 - 122153082 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437762_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (31)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009735.3 NP_033865.2 1 - 2 111 77
          GenBank mRNA AK019389.1 - 1 - 2 114 77
          AK028378.1 - 1 - 2 113 77
          AK153255.1 - 1 - 2 113 77
          AK166067.1 - 1 - 2 114 77
          BC085164.1 - 1 - 2 66 6846389 77
          X01838.1 - 1 - 2 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001838.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Endogenous Controls Recommended Assay Mus musculus Mouse Endogenous Controls Inventoried Mouse_Endogenous_Controls Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B2m

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          immune system process
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          cellular defense response
          T cell differentiation in thymus
          protein refolding
          iron ion homeostasis
          negative regulation of receptor binding
          glycoprotein binding
          protein binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200014262.mm.1 NM_009735.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014262.mm.1 NM_009735.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014262.mm.1 TC0200001838.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014267.mm.1 NM_009735.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014267.mm.1 NM_009735.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014267.mm.1 TC0200001838.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014264.mm.1 NM_009735.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014264.mm.1 NM_009735.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014264.mm.1 TC0200001838.mm.1 MTA TC

          Back To Top
           

          Citations (31)

          Gene expression profiling in the lung and liver of PFOA-exposed mouse fetuses.
          Authors
          Rosen MB, Thibodeaux JR, Wood CR, Zehr RD, Schmid JE, Lau C
          Journal
          Toxicology - Page No 15 (Mon Sep 24 00:00:00 UTC 2007)
          Volume (Year)
          239 (2007)
          Product Usage
          -
          Calorie restriction increases fatty acid synthesis and whole body fat oxidation rates.
          Authors
          Bruss MD, Khambatta CF, Ruby MA, Aggarwal I, Hellerstein MK
          Journal
          Am J Physiol Endocrinol Metab - Page No E108 (Fri Jan 01 00:00:00 UTC 2010)
          Volume (Year)
          298 (2010)
          Product Usage
          -
          Plasticity of proximal-distal cell fate in the mammalian limb bud.
          Authors
          Wyngaarden LA, Hopyan S
          Journal
          Dev Biol - Page No 225 (Tue Jan 01 00:00:00 UTC 2008)
          Volume (Year)
          313 (2008)
          Product Usage
          -
          Interleukin-6 trans-signaling regulates glycogen consumption after D-galactosamine-induced liver damage.
          Authors
          Drucker C, Rabe B, Chalaris A, Schulz E, Scheller J, Rose-John S
          Journal
          J Interferon Cytokine Res - Page No 711 (Sun Nov 01 00:00:00 UTC 2009)
          Volume (Year)
          29 (2009)
          Product Usage
          -
          Transcriptomic comparison of the retina in two mouse models of diabetes.
          Authors
          Freeman WM, Bixler GV, Brucklacher RM, Walsh E, Kimball SR, Jefferson LS, Bronson SK
          Journal
          J Ocul Biol Dis Infor - Page No 202 (Thu Jan 01 00:00:00 UTC 2009)
          Volume (Year)
          2 (2009)
          Product Usage
          -
          Effects of PI3K catalytic subunit and Akt isoform deficiency on mTOR and p70S6K activation in myoblasts.
          Authors
          Matheny RW Jr, Adamo ML
          Journal
          Biochem Biophys Res Commun - Page No 252 (Fri Dec 11 00:00:00 UTC 2009)
          Volume (Year)
          390 (2009)
          Product Usage
          -
          Gene expression profiling in the liver and lung of perfluorooctane sulfonate-exposed mouse fetuses: comparison to changes induced by exposure to perfluorooctanoic acid.
          Authors
          Rosen MB, Schmid JE, Das KP, Wood CR, Zehr RD, Lau C
          Journal
          Reprod Toxicol - Page No 278 (Mon Jun 01 00:00:00 UTC 2009)
          Volume (Year)
          27 (2009)
          Product Usage
          -
          Growth hormone-releasing peptide-2 stimulates secretion and synthesis of adrenocorticotropic hormone in mouse pituitary.
          Authors
          Kageyama K, Kushibiki M, Hanada K, Sakihara S, Yasujima M, Suda T
          Journal
          Regul Pept - Page No 116 (Fri Nov 27 00:00:00 UTC 2009)
          Volume (Year)
          158 (2009)
          Product Usage
          -
          Differential expression of interferon (IFN) regulatory factors and IFN-stimulated genes at early times after West Nile virus infection of mouse embryo fibroblasts.
          Authors
          Scherbik SV, Stockman BM, Brinton MA
          Journal
          J Virol - Page No 12005 (Thu Nov 01 00:00:00 UTC 2007)
          Volume (Year)
          81 (2007)
          Product Usage
          -
          Gender-dependent expression of murine Irf5 gene: implications for sex bias in autoimmunity.
          Authors
          Shen H, Panchanathan R, Rajavelu P, Duan X, Gould KA, Choubey D
          Journal
          J Mol Cell Biol - Page No 284 (Fri Oct 01 00:00:00 UTC 2010)
          Volume (Year)
          2 (2010)
          Product Usage
          -
          Mouse embryo fibroblasts lacking the tumor suppressor menin show altered expression of extracellular matrix protein genes.
          Authors
          Ji Y, Prasad NB, Novotny EA, Kaur S, Elkahloun A, Chen Y, Zhang RZ, Chu ML, Agarwal SK, Marx SJ, Collins FS, Chandrasekharappa SC
          Journal
          Mol Cancer Res - Page No 1041 (Mon Oct 01 00:00:00 UTC 2007)
          Volume (Year)
          5 (2007)
          Product Usage
          -
          Genetic control of resistance to hepatocarcinogenesis by the mouse Hpcr3 locus.
          Authors
          Manenti G, Galvan A, Falvella FS, Pascale RM, Spada E, Milani S, Gonzalez Neira A, Feo F, Dragani TA
          Journal
          Hepatology - Page No 617 (Fri Aug 01 00:00:00 UTC 2008)
          Volume (Year)
          48 (2008)
          Product Usage
          -
          Stimulation of osteoblasts with Emdogain increases the expression of specific mineralization markers.
          Authors
          Weishaupt P, Bernimoulin JP, Trackman P, Hagewald S
          Journal
          Oral Surg Oral Med Oral Pathol Oral Radiol Endod - Page No 304 (Fri Aug 01 00:00:00 UTC 2008)
          Volume (Year)
          106 (2008)
          Product Usage
          -
          Early organ-specific endothelial activation during hemorrhagic shock and resuscitation.
          Authors
          van Meurs M, Wulfert FM, Knol AJ, De Haes A, Houwertjes M, Aarts LP, Molema G
          Journal
          Shock - Page No 291 (Fri Feb 01 00:00:00 UTC 2008)
          Volume (Year)
          29 (2008)
          Product Usage
          -
          Molecular structure of tail tendon fibers in TIEG1 knockout mice using synchrotron diffraction technology.
          Authors
          Gumez L, Bensamoun SF, Doucet J, Haddad O, Hawse JR, Subramaniam M, Spelsberg TC, Pichon C
          Journal
          J Appl Physiol - Page No 1706 (Tue Jun 01 00:00:00 UTC 2010)
          Volume (Year)
          108 (2010)
          Product Usage
          -
          Selection of reference genes for quantitative real-time RT-PCR studies in mouse brain.
          Authors
          Boda E, Pini A, Hoxha E, Parolisi R, Tempia F
          Journal
          J Mol Neurosci - Page No 238 (Sun Mar 01 00:00:00 UTC 2009)
          Volume (Year)
          37 (2009)
          Product Usage
          -
          PDGF-Ralpha gene expression predicts proliferation, but PDGF-A suppresses transdifferentiation of neonatal mouse lung myofibroblasts.
          Authors
          Kimani PW, Holmes AJ, Grossmann RE, McGowan SE
          Journal
          Respir Res - Page No 119 (Thu Jan 01 00:00:00 UTC 2009)
          Volume (Year)
          10 (2009)
          Product Usage
          -
          Gene expression fluctuations in murine hematopoietic stem cells with cell cycle progression.
          Authors
          Dooner GJ, Colvin GA, Dooner MS, Johnson KW, Quesenberry PJ
          Journal
          J Cell Physiol - Page No 786 (Sat Mar 01 00:00:00 UTC 2008)
          Volume (Year)
          214 (2008)
          Product Usage
          -
          The matricellular protein periostin is required for sost inhibition and the anabolic response to mechanical loading and physical activity.
          Authors
          Bonnet N, Standley KN, Bianchi EN, Stadelmann V, Foti M, Conway SJ, Ferrari SL
          Journal
          J Biol Chem - Page No 35939 (Fri Dec 18 00:00:00 UTC 2009)
          Volume (Year)
          284 (2009)
          Product Usage
          -
          Lysyl oxidase (lox) gene deficiency affects osteoblastic phenotype.
          Authors
          Pischon N, Maki JM, Weisshaupt P, Heng N, Palamakumbura AH, N'Guessan P, Ding A, Radlanski R, Renz H, Bronckers TA, Myllyharju J, Kielbassa AM, Kleber BM, Bernimoulin JP, Trackman PC
          Journal
          Calcif Tissue Int - Page No 119 (Sat Aug 01 00:00:00 UTC 2009)
          Volume (Year)
          85 (2009)
          Product Usage
          -
          Adiponectin suppresses tumorigenesis in Apc(Min)(/+) mice.
          Authors
          Otani K, Kitayama J, Yasuda K, Nio Y, Iwabu M, Okudaira S, Aoki J, Yamauchi T, Kadowaki T, Nagawa H
          Journal
          Cancer Lett - Page No 177 (Sun Feb 28 00:00:00 UTC 2010)
          Volume (Year)
          288 (2010)
          Product Usage
          -
          Type I interferons produced by resident renal cells may promote end-organ disease in autoantibody-mediated glomerulonephritis.
          Authors
          Fairhurst AM, Xie C, Fu Y, Wang A, Boudreaux C, Zhou XJ, Cibotti R, Coyle A, Connolly JE, Wakeland EK, Mohan C
          Journal
          J Immunol - Page No 6831 (Sun Nov 15 00:00:00 UTC 2009)
          Volume (Year)
          183 (2009)
          Product Usage
          -
          Cyanidin 3-glucoside ameliorates hyperglycemia and insulin sensitivity due to downregulation of retinol binding protein 4 expression in diabetic mice.
          Authors
          Sasaki R, Nishimura N, Hoshino H, Isa Y, Kadowaki M, Ichi T, Tanaka A, Nishiumi S, Fukuda I, Ashida H, Horio F, Tsuda T
          Journal
          Biochem Pharmacol - Page No 1619 (Mon Dec 03 00:00:00 UTC 2007)
          Volume (Year)
          74 (2007)
          Product Usage
          -
          Non-proteolytic effect of beta-site APP-cleaving enzyme 1 (BACE1) on sodium channel function.
          Authors
          Huth T, Schmidt-Neuenfeldt K, Rittger A, Saftig P, Reiss K, Alzheimer C
          Journal
          Neurobiol Dis - Page No 282 (Sun Feb 01 00:00:00 UTC 2009)
          Volume (Year)
          33 (2009)
          Product Usage
          -
          Invariant natural killer T cell-natural killer cell interactions dictate transplantation outcome after alpha-galactosylceramide administration.
          Authors
          Kuns RD, Morris ES, Macdonald KP, Markey KA, Morris HM, Raffelt NC, Banovic T, Don AL, Rowe V, Burman AC, Clouston AD, Farah C, Besra GS, Illarionov PA, Smyth MJ, Porcelli SA, Hill GR
          Journal
          Blood - Page No 5999 (Thu Jun 04 00:00:00 UTC 2009)
          Volume (Year)
          113 (2009)
          Product Usage
          -
          Microvesicle entry into marrow cells mediates tissue-specific changes in mRNA by direct delivery of mRNA and induction of transcription.
          Authors
          Aliotta JM, Pereira M, Johnson KW, de Paz N, Dooner MS, Puente N, Ayala C, Brilliant K, Berz D, Lee D, Ramratnam B, McMillan PN, Hixson DC, Josic D, Quesenberry PJ
          Journal
          Exp Hematol - Page No 233 (Mon Mar 01 00:00:00 UTC 2010)
          Volume (Year)
          38 (2010)
          Product Usage
          -
          Activation of antioxidative enzymes induced by low-dose-rate whole-body gamma irradiation: adaptive response in terms of initial DNA damage.
          Authors
          Otsuka K, Koana T, Tauchi H, Sakai K
          Journal
          Radiat Res - Page No 474 (Fri Sep 01 00:00:00 UTC 2006)
          Volume (Year)
          166 (2006)
          Product Usage
          -
          Involvement of nuclear factor-kB and Nurr-1 in cytokine-induced transcription of proopiomelanocortin gene in AtT20 corticotroph cells.
          Authors
          Takayasu S, Iwasaki Y, Nigawara T, Asai M, Yoshida M, Kageyama K, Suda T
          Journal
          Neuroimmunomodulation - Page No 88 (Fri Jan 01 00:00:00 UTC 2010)
          Volume (Year)
          17 (2010)
          Product Usage
          -
          Histone deacetylase inhibition reduces myocardial ischemia-reperfusion injury in mice.
          Authors
          Granger A, Abdullah I, Huebner F, Stout A, Wang T, Huebner T, Epstein JA, Gruber PJ
          Journal
          Faseb J - Page No 3549 (Wed Oct 01 00:00:00 UTC 2008)
          Volume (Year)
          22 (2008)
          Product Usage
          -
          PI3K p110 alpha and p110 beta have differential effects on Akt activation and protection against oxidative stress-induced apoptosis in myoblasts.
          Authors
          Matheny RW Jr, Adamo ML
          Journal
          Cell Death Differ - Page No 677 (Thu Apr 01 00:00:00 UTC 2010)
          Volume (Year)
          17 (2010)
          Product Usage
          -
          Dehydroepiandrosterone modulates toll-like receptor expression on splenic macrophages of mice after severe polymicrobial sepsis.
          Authors
          Matsuda A, Furukawa K, Suzuki H, Matsutani T, Tajiri T, Chaudry IH
          Journal
          Shock - Page No 364 (Sat Oct 01 00:00:00 UTC 2005)
          Volume (Year)
          24 (2005)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline