Search
Search

Bub1b| Gene Symbol: Bub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009773.3 | NP_033903.2 | 18 - 19 | 2588 | 98 | |
| GenBank mRNA | AF107296.1 | - | 18 - 19 | 2543 | 98 | |
| AK088502.1 | - | 4 - 5 | 644 | 98 | ||
| AK133909.1 | - | 18 - 19 | 2529 | 98 | ||
| BC031577.1 | - | 18 - 19 | 2539 | 4953223 | 98 | |
Set Membership: |
|