Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438057_g1
          See other SERPINH1 GE Assays ›
          Gene Symbol:
          Serpinh1
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade H, member 1
          Gene Aliases:
          BERF-1, Cbp1, Cbp2, Hsp47, J6, Serpinh2, gp46
          Chromosome Location:
          Chr.7: 99345375 - 99353239 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438057_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001111043.1 NP_001104513.1 4 - 5 989 71
          NM_001111044.1 NP_001104514.1 3 - 4 945 71
          NM_001285776.1 NP_001272705.1 4 - 5 1029 71
          NM_009825.2 NP_033955.2 3 - 4 1116 71
          GenBank mRNA AK029080.1 - 4 - 5 907 71
          AK031457.1 - 3 - 4 994 71
          AK076474.1 - 3 - 4 71
          AK077660.1 - 3 - 4 822 71
          AK077937.1 - 3 - 4 848 71
          AK079460.1 - 3 - 4 821 71
          AK086519.1 - 3 - 4 821 71
          AK086523.1 - 3 - 4 821 71
          AK089993.1 - 3 - 4 823 71
          AK090103.1 - 3 - 4 823 71
          AK159697.1 - 3 - 4 823 71
          AK159998.1 - 3 - 4 823 71
          AK167403.1 - 3 - 4 821 71
          AK168442.1 - 3 - 4 824 71
          AK168870.1 - 3 - 4 799 71
          BC085143.1 - 4 - 5 862 6415383 71
          J05609.1 - 4 - 5 856 71
          X60676.1 - 3 - 4 971 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003797.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinh1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          chondrocyte development involved in endochondral bone morphogenesis
          negative regulation of endopeptidase activity
          collagen fibril organization
          collagen biosynthetic process
          protein maturation
          serine-type endopeptidase inhibitor activity
          protein binding
          collagen binding
          poly(A) RNA binding
          unfolded protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_001111043.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_001111043.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_001111044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_001111044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_001285776.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_009825.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 NM_009825.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031969.mm.1 TC0700003797.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_001111043.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_001111043.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_001111044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_001111044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_001285776.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_009825.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 NM_009825.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031968.mm.1 TC0700003797.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline