Search
Search

Fgf14| Gene Symbol: Fgf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010201.4 | NP_034331.2 | 2 - 3 | 329 | 86 | |
| NM_207667.3 | NP_997550.1 | 2 - 3 | 729 | 86 | ||
| GenBank mRNA | AB008907.1 | - | 2 - 3 | 319 | 86 | |
| AB029498.1 | - | 1 - 2 | 52 | 86 | ||
| AK030616.1 | - | 2 - 3 | 688 | 86 | ||
| AK035974.1 | - | 2 - 3 | 339 | 86 | ||
| AK079100.1 | - | 2 - 3 | 729 | 86 | ||
| BC107003.1 | - | 3 - 4 | 663 | 40046696 | 86 | |
| BC107004.1 | - | 2 - 3 | 623 | 40046697 | 86 | |
| U66204.1 | - | 2 - 3 | 304 | 86 | ||
Set Membership: |
|