Search
Search

Gabra1| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010250.5 | NP_034380.1 | 7 - 8 | 1477 | 74 | |
| GenBank mRNA | AK034095.1 | - | 7 - 8 | 1478 | 74 | |
| AK039496.1 | - | 7 - 8 | 761 | 74 | ||
| AK141596.1 | - | 7 - 8 | 1337 | 74 | ||
| BC132331.1 | - | 7 - 8 | 1153 | 40130608 | 74 | |
| BC137733.1 | - | 7 - 8 | 1156 | 8860752 | 74 | |
| M63436.1 | - | 7 - 8 | 1150 | 74 | ||
| M86566.1 | - | 7 - 8 | 845 | 74 | ||
| X61430.1 | - | 6 - 7 | 733 | 74 | ||
Set Membership: |
|