Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439116_s1
          See other GJA10 GE Assays ›
          Gene Symbol:
          Gja10
          Gene Name:
          gap junction protein, alpha 10
          Gene Aliases:
          Cx-57, Cx59, cx57
          Chromosome Location:
          Chr.4: 32600865 - 32602382 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439116_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gja10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010289.2 NP_034419.2 1 1056 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002364.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gja10

          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s) Process(es)

          cell communication
          gamete generation
          synapse assembly
          response to light stimulus
          detection of light stimulus involved in visual perception
          transmembrane transport
          gap junction channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143341 NM_010289.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169464 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143341 NM_010289.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22822398 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169464 - -
          Whole Mouse Genome G4122A A_51_P130940 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120136 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169464 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03337212 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03337221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03337220 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03337216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22822417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04090481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03657661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04090479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03658532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22822394 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10511900 - -
          Affymetrix HT MG-430 PM Array Plate 1422328_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10511898 - -
          Mouse Genome 430 2.0 Array 1422328_PM_at - -
          Mouse Genome 430 2.0 Array 1422328_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14610_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14610_at - -
          Affymetrix Mouse Clariom S Array PSR0400019211.mm.1 NM_010289.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019211.mm.1 NM_010289.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019211.mm.1 TC0400002364.mm.1 MTA TC
          Mouse Expression Set 430 1422328_at - -
          Mouse Expression Set 430 1422328_PM_at - -
          Murine Genome U74v2 Set 102164_at - -
          Murine Genome U74v2 Set 102164_PM_at - -
          Mouse Genome 430A 2.0 Array 1422328_at - -
          Mouse Genome 430A 2.0 Array 1422328_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422328_at - -
          Affymetrix HT Mouse Genome 430A Array 1422328_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664913 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060463 - -
          Illumina mouse-6 v1.0 expression beadchip 2060463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717340 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717340 - -
          mouse-6 v1.1 (Illumina) scl2495.1.1_145-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl2495.1.1_145-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717340 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline