Search
Search

Ifnar1| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010508.2 | NP_034638.2 | 3 - 4 | 508 | 109 | |
| GenBank mRNA | AK132431.1 | - | 3 - 4 | 440 | 109 | |
| AK136950.1 | - | 3 - 4 | 485 | 109 | ||
| AK141630.1 | - | 3 - 4 | 444 | 109 | ||
| AK148246.1 | - | 3 - 4 | 450 | 109 | ||
| AK157555.1 | - | 3 - 4 | 475 | 109 | ||
| BC043935.1 | - | 3 - 4 | 459 | 4507009 | 109 | |
| BC052217.1 | - | 3 - 4 | 468 | 30008497 | 109 | |
| BC052429.1 | - | 3 - 4 | 470 | 6836300 | 109 | |
| M89641.1 | - | 3 - 4 | 473 | 109 | ||
Set Membership: |
|