Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439646_m1
          See other IL5 GE Assays ›
          Gene Symbol:
          Il5
          Gene Name:
          interleukin 5
          Gene Aliases:
          Il-5
          Chromosome Location:
          Chr.11: 53719364 - 53725623 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439646_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010558.1 NP_034688.1 2 - 3 219 62
          GenBank mRNA BC125366.1 - 2 - 3 204 40129881 62
          BC132066.1 - 2 - 3 204 40130343 62
          X04601.1 - 2 - 3 219 62
          X06270.1 - 2 - 3 219 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000612.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus Mouse Chemokine Panel Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il5

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          positive regulation of cell proliferation
          cytokine-mediated signaling pathway
          positive regulation of B cell proliferation
          positive regulation of eosinophil differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of JAK-STAT cascade
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of immunoglobulin secretion
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of podosome assembly
          cytokine activity
          interleukin-5 receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138386 X04601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138386 X06270.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138386 NM_010558.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153191 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140600 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138386 X04601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138386 X06270.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138386 NM_010558.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185415 X04601.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185415 X06270.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185415 NM_010558.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27806267 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140600 - -
          Whole Mouse Genome G4122A A_51_P185415 X04601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185415 X06270.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185415 NM_010558.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120892 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140600 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09102581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09648978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09648977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09102580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09102578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27806311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09101515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09121057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09438582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09102580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09031699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09201899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09556165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09498749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09189018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09490605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376059 - -
          Affymetrix HT MG-430 PM Array Plate 1450550_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376054 - -
          Mouse Genome 430 2.0 Array 1450550_PM_at - -
          Mouse Genome 430 2.0 Array 1450550_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16191_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16191_at - -
          Affymetrix Mouse Clariom S Array PSR1100004864.mm.1 NM_010558.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004864.mm.1 NM_010558.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004864.mm.1 TC1100000612.mm.1 MTA TC
          Mouse Expression Set 430 1450550_PM_at - -
          Mouse Expression Set 430 1450550_at - -
          Murine Genome U74v2 Set 98374_at - -
          Murine Genome U74v2 Set 98374_PM_at - -
          Mouse Genome 430A 2.0 Array 1450550_PM_at - -
          Mouse Genome 430A 2.0 Array 1450550_at - -
          Affymetrix HT Mouse Genome 430A Array 1450550_at - -
          Affymetrix HT Mouse Genome 430A Array 1450550_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 335478 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6370364 - -
          Illumina mouse-6 v1.0 expression beadchip 6370364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731160 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731160 - -
          mouse-6 v1.1 (Illumina) scl41561.4.502_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370364 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41561.4.502_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731160 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline