Search
Search

Itga4| Gene Symbol: Itga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010576.3 | NP_034706.3 | 6 - 7 | 986 | 122 | |
| GenBank mRNA | AB850880.1 | - | 6 - 7 | 751 | 122 | |
| AF109136.1 | - | 6 - 7 | 822 | 122 | ||
| AK051676.1 | - | 6 - 7 | 912 | 122 | ||
| AK078842.1 | - | 6 - 7 | 691 | 122 | ||
| AK089898.1 | - | 5 - 6 | 786 | 122 | ||
| AK156015.1 | - | 6 - 7 | 882 | 122 | ||
| AK156517.1 | - | 6 - 7 | 756 | 122 | ||
| BC068313.1 | - | 6 - 7 | 766 | 30093248 | 122 | |
| U97151.1 | - | 4 - 5 | 354 | 122 | ||
| X53176.1 | - | 6 - 7 | 909 | 122 | ||
Set Membership: |
|