Search
Search
| Gene Symbol: Lcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010695.1 | NP_034825.1 | 2 - 3 | 235 | 85 | |
| GenBank mRNA | BC114544.1 | - | 2 - 3 | 234 | 40050061 | 85 |
| D38581.1 | - | 2 - 3 | 235 | 85 | ||
Set Membership: |
|