Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440855_m1
          See other PRKCA GE Assays ›
          Gene Symbol:
          Prkca
          Gene Name:
          protein kinase C, alpha
          Gene Aliases:
          AI875142, Pkca
          Chromosome Location:
          Chr.11: 107933387 - 108343888 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440855_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkca
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011101.3 NP_035231.2 13 - 14 1743 72
          GenBank mRNA AK147084.1 - 13 - 14 1744 72
          AK163950.1 - 3 - 4 431 72
          BC096493.1 - 13 - 14 1537 30742571 72
          M25811.1 - 13 - 14 1777 72
          X52684.1 - 14 - 15 1684 72
          X52685.1 - 14 - 15 1684 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003966.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkca

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          inactivation of MAPK activity
          response to reactive oxygen species
          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          regulation of the force of heart contraction
          chondrocyte differentiation
          desmosome assembly
          protein phosphorylation
          negative regulation of protein kinase activity
          cellular calcium ion homeostasis
          apoptotic process
          regulation of muscle contraction
          cell adhesion
          learning or memory
          negative regulation of cell proliferation
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          phosphorylation
          negative regulation of translation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          central nervous system neuron axonogenesis
          positive regulation of cell migration
          neutrophil chemotaxis
          positive regulation of lipopolysaccharide-mediated signaling pathway
          response to estradiol
          negative regulation of glial cell apoptotic process
          histone H3-T6 phosphorylation
          intracellular signal transduction
          peptidyl-serine autophosphorylation
          positive regulation of blood vessel endothelial cell migration
          response to ethanol
          positive regulation of macrophage differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          negative regulation of heart contraction
          positive regulation of exocytosis
          positive regulation of mitotic cell cycle
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          protein autophosphorylation
          regulation of receptor-mediated endocytosis
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          regulation of peptidyl-tyrosine phosphorylation
          induction of positive chemotaxis
          positive regulation of synapse assembly
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          cellular response to carbohydrate stimulus
          regulation of platelet aggregation
          intrinsic apoptotic signaling pathway
          positive regulation of dense core granule biogenesis
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-dependent protein kinase C activity
          protein binding
          ATP binding
          zinc ion binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          histone kinase activity (H3-T6 specific)
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100037222.mm.1 NM_011101.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037222.mm.1 NM_011101.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037222.mm.1 TC1100003966.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100037223.mm.1 NM_011101.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037223.mm.1 NM_011101.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037223.mm.1 TC1100003966.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline