Search
Search

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011104.3 | NP_035234.1 | 6 - 7 | 1624 | 94 | |
| GenBank mRNA | AF028009.1 | - | 6 - 7 | 825 | 94 | |
| AF325507.1 | - | 6 - 7 | 816 | 94 | ||
| AK082438.1 | - | 6 - 7 | 1625 | 94 | ||
| AK085351.1 | - | 7 - 8 | 1036 | 94 | ||
Set Membership: |
|